missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPZ-015666, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | EPZ-015666 |
| CAS | 1616391-65-1 |
| Molecular Formula | C20H25N5O3 |
| Quantity | 1 mg |
| Synonym | N-[(2S)-3-(3,4-Dihydro-2(1H)-isoquinolinyl)-2-hydroxypropyl]-6-(3-oxetanylamino)-4-pyrimidinecarboxamide |
| InChI Formula | InChI=1S/C20H25N5O3/c26-17(10-25-6-5-14-3-1-2-4-15(14)9-25)8-21-20(27)18-7-19(23-13-22-18)24-16-11-28-12-16/h1-4,7,13,16-17,26H,5-6,8-12H2,(H,21,27)(H,22,23,24)/t17-/m0/s1 |
| SMILES | O[C@@H](CNC(C1=NC=NC(NC2COC2)=C1)=O)CN3CC4=CC=CC=C4CC3 |
| IUPAC Name | (S)-N-(3-(3,4-dihydroisoquinolin-2(1H)-yl)-2-hydroxypropyl)-6-(oxetan-3-ylamino)pyrimidine-4-carboxamide |
| Molecular Weight (g/mol) | 383.44 |
| Formula Weight | 383.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction