missing translation for 'onlineSavingsMsg'
Learn More

EPZ015666, MedChemExpress

Product Code. 16491507
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491507

Brand: Medchem Express HY12727

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 with an IC50 of 22 nM.

EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 with an IC50 of 22 nM.

Specifications

Chemical Name or Material EPZ015666
CAS 1616391-65-1
Color White
Purity Grade Notes Research
Molecular Formula C20H25N5O3
Quantity 50 mg
Synonym GSK3235025
Solubility Information DMSO : 100 mg/mL (260.80 mM; Need ultrasonic)
SMILES O=C(NC[C@H](O)CN1CCC(C=CC=C2)=C2C1)C3=CC(NC4COC4)=NC=N3
Molecular Weight (g/mol) 383.44
Formula Weight 383.44
Percent Purity 98.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.