missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EQTN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553312
This item is not returnable.
View return policy
Description
Immunogen sequence: EDVNKQEEKN EDHTPNYAPA NEKNGNYYKD IKQYVFTTQN PNGTESEISV RATTDLNFAL KNGSTPNVPA FWTMLAKAIN GTAVVMDDKD QLFHPIPESD VNATQGENQP DLEDLKIK Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.
Specifications
| EQTN | |
| Polyclonal | |
| Unconjugated | |
| EQTN | |
| acrosome formation associated factor; acrosome formation-associated factor; AFAF; C9orf11; EQTN; Equatorin; equatorin, sperm acrosome associated; SPACA8; sperm acrosome associated 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQ60 | |
| EQTN | |
| Recombinant protein corresponding to Human EQTN. Recombinant protein control fragment (Product #RP-90155). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction