missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Equine IL-10 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF1605
This item is not returnable.
View return policy
Description
IL-10 Polyclonal specifically detects IL-10 in Equine samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| IL-10 | |
| Polyclonal | |
| Biotin | |
| Q28374 | |
| IL10 | |
| E. coli-derived recombinant equine IL-10 Ser19-Asn178 Accession # Q28374 | |
| 50 μg | |
| Primary | |
| Detects equine IL-10 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant canine IL-10 and recombinant porcine IL-10, and less than 0.4% cross-reactivity with recombinant human IL-10, recombinant mouse IL-10, recombinant rat IL-10, and recombinant feline IL-10 is observed. | |
| Equine | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3586 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction