missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Equine IL-1ra/IL-1F3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2466-SP
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Polyclonal specifically detects IL-1ra/IL-1F3/IL1RN in Equine samples. It is validated for Western Blot, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3557 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| O18999 | |
| IL1RN | |
| E. coli-derived recombinant equine IL-1ra/IL-1F3 His26-Gln177 Accession # O18999 | |
| 25 μg | |
| Primary | |
| Detects equine IL-1ra/IL-1F3 in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant mouse IL-1ra is observed, less than 5% cross-reactivity with recombinant rat IL-1ra is observed, less than 2% cross-reactivity with recombinant porcine IL-1ra is observed, less than 0.4% cross-reactivity with recombinant human IL-1ra is observed, and less than 0.2% cross-reactivity with recombinant equine IL-1beta is observed. | |
| Equine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction