missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Equine IL-5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2470
This item is not returnable.
View return policy
Description
IL-5 Polyclonal specifically detects IL-5 in Equine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| IL-5 | |
| Polyclonal | |
| Unconjugated | |
| O02699 | |
| IL5 | |
| Mouse myeloma cell line NS0-derived recombinant equine IL-5 Leu20-Gly134 Accession # O02699 | |
| 100 μg | |
| Primary | |
| Detects equine IL-5 in direct ELISAs and Western blots. | |
| Equine | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.500-6.00 ug/mL, Immunocytochemistry 10-25 ug/mL | |
| BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3567 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction