missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER alpha/NR3A1 Antibody (33), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
€ 213.00
Specifications
| Antigen | ER alpha/NR3A1 |
|---|---|
| Clone | 33 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 4 μg/mL, Immunohistochemistry 1:20 - 1:100, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunofluorescence 10 μg/mL |
| Applications | Western Blot |
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human, Mouse, Rat, Avian, Bovine, Gerbil, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
| ER alpha/NR3A1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| P03372 | |
| 2099 | |
| Synthetic peptide corresponding to the residues E(247) V G M M K G G I R K D R R G(261) of the ER alpha DNA binding domain. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 67 kDa |
| 33 | |
| Western Blot 4 μg/mL, Immunohistochemistry 1:20 - 1:100, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunofluorescence 10 μg/mL | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| ESR1 | |
| IgG1 | |
| Protein G purified | |
| Detects both the steroid occupied and unoccupied forms of the estrogen receptor (ER) alpha. This does not cross react with ER beta, androgen receptor, progesterone receptor, or glucocorticoid receptor. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title