missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER alpha/NR3A1 Antibody (ER505), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 551.00 - € 594.00
Specifications
| Antigen | ER alpha/NR3A1 |
|---|---|
| Clone | ER505 |
| Dilution | Western Blot |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18468381
|
Novus Biologicals
NBP2-33106APC-100ul |
100 μL |
€ 594.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18438011
|
Novus Biologicals
NBP2-33106APC |
0.1 mL |
€ 551.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ER alpha/NR3A1 Monoclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western BlotSpecifications
| ER alpha/NR3A1 | |
| Western Blot | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| Recombinant human ER alpha/NR3A1 alpha protein (aa2-185) (Uniprot: P03372) | |
| Primary | |
| Store at 4C in the dark. |
| ER505 | |
| Western Blot | |
| APC | |
| Mouse | |
| Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| PBS | |
| 2099 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title