missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER beta/NR3A2 Antibody (PPG5/25), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61942B
This item is not returnable.
View return policy
Description
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ER beta/NR3A2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ESR2 | |
| Tuberculin conjugated peptide that recognizes C terminus of ER beta5 at sequence LLSHVRHARYAP | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| PPG5/25 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
| Mouse | |
| Protein A purified | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 2100 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction