missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYPB Antibody (OTI1E1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71850AF532
This item is not returnable.
View return policy
Description
HYPB Monoclonal specifically detects HYPB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HYPB | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SETD2 | |
| Recombinant protein fragment corresponding to amino acids 1787-2144 of human HYPB (NP_054878) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1E1 | |
| Western Blot, Immunohistochemistry | |
| FLJ22472, FLJ23184, FLJ45883, FLJ46217, HIF1, HIF-1EC 2.1.1.43, HIP-1, histone-lysine N-methyltransferase SETD2, hSET2, huntingtin interacting protein 1, Huntingtin yeast partner B, Huntingtin-interacting protein 1, Huntingtin-interacting protein B, HYPBp231HBP, KIAA1732SET domain-containing protein 2, KMT3AHBP231, Lysine N-methyltransferase 3A, SET domain containing 2, SET2FLJ16420 | |
| Mouse | |
| 0.1 mL | |
| 29072 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction