missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555073
This item is not returnable.
View return policy
Description
Immunogen sequence: SVLVMNKVDC LKQKSVLLEL TAALTEGVVN GKKLKMRQAF HSHPGTHCPS PAVKDPNTQS VGNPQRIGWP HFKEIFMLSA LSQEDVKTLK QY Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.
The protein encoded by this gene is a GTPase that localizes to the mitochondrion. The encoded protein binds to the 3' terminal stem loop of 12S mitochondrial rRNA and is required for proper assembly of the 28S small mitochondrial ribosomal subunit. Deletion of this gene has been shown to cause mitochondrial dysfunction, growth retardation, and apoptosis. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| ERAL1 | |
| Polyclonal | |
| Unconjugated | |
| ERAL1 | |
| 2610524P08Rik; 9130407C09Rik; AU019798; CEGA; Conserved ERA-like GTPase; ERA; era (E. coli G-protein homolog)-like 1; Era (G-protein)-like 1; Era (G-protein)-like 1 (E. coli); Era G-protein-like 1; Era like 12S mitochondrial rRNA chaperone 1; Eral1; ERAL1A; Era-like 1; Era-like 12S mitochondrial rRNA chaperone 1; ERA-like GTPase; ERA-like protein 1; ERA-W; GTPase Era, mitochondrial; GTPase, human homolog of E. coli essential cell cycle protein Era; GTP-binding protein era homolog; HERA; H-ERA; HERA-A; HERA-B; Mera; M-ERA; MERA-S; MERA-W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26284 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75616 | |
| ERAL1 | |
| Recombinant protein corresponding to Human ERAL1. Recombinant protein control fragment (Product #RP-93314). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction