missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERAP2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547541
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Aminopeptidases hydrolyze N-terminal amino acids of proteins or peptide substrates. Major histocompatibility complex class I molecules rely on aminopeptidases such as ERAP1 and LRAP to trim precursors to antigenic peptides in the endoplasmic reticulum following cleavage in the cytoplasm by tripeptidyl peptidase II.
Specifications
| ERAP2 | |
| Polyclonal | |
| Unconjugated | |
| ERAP2 | |
| Endoplasmic reticulum aminopeptidase 2; ERAP2; FLJ23633; FLJ23701; FLJ23807; Leukocyte-derived arginine aminopeptidase; LRAP; L-RAP | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64167 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q6P179 | |
| ERAP2 | |
| Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase LRAP/ERAP2 Ala56-Thr960. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction