missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Antibody (HRB2/258), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34641AF700
This item is not returnable.
View return policy
Description
ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ErbB2/Her2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| Mouse | |
| 185 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| 2064 | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| HRB2/258 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| ERBB2 | |
| Recombinant extracellular domain of human ErbB2/Her2 protein (Uniprot: IDP04626) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 185kDa, which is identified as c-erbB-2/HER-2/neu. Its epitope is localized in the extracellular domain. C-erbB-2/HER-2 is a member of the EGFR family. This monoclonal antibody is specific and shows minimal cross-reaction with other members of the EGFR-family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. C-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction