missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Mouse, Clone: HRB2/258, Novus Biologicals™
Mouse Monoclonal Antibody
€ 290.00 - € 509.00
Specifications
| Antigen | ErbB2/Her2 |
|---|---|
| Clone | HRB2/258 |
| Dilution | Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18185013
|
Novus Biologicals
NBP2-32862-0.02MG |
0.02 mg |
€ 290.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18127434
|
Novus Biologicals
NBP2-32862-0.1MG |
0.1 mg |
€ 509.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ErbB2/Her2 | |
| Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| P04626 | |
| 2064 | |
| Recombinant extracellular domain of human HER-2 protein | |
| Primary | |
| Store at 4C. | |
| 185 kDa |
| HRB2/258 | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| ERBB2 | |
| IgG2a κ | |
| Protein A purified | |
| Recognizes a protein of 185kDa, which is identified as c-erbB-2/HER-2/neu. Its epitope is localized in the extracellular domain. C-erbB-2/HER-2 is a member of the EGFR family. This MAb is specific and shows minimal cross-reaction with other members of the EGFR-family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. C-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title