missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERBB2 Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505807S
This item is not returnable.
View return policy
Description
ERBB2 Monoclonal antibody specifically detects ERBB2 in Canine, Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ERBB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P46734 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 489547, 5606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G7 | |
| Unconjugated | |
| MAP2K3 | |
| avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb | |
| Full length human recombinant protein of human MAP2K3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction