missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ErbB3/Her3 Antibody (seribantumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28218-100ug
This item is not returnable.
View return policy
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ErbB3/Her3 | |
| Monoclonal | |
| Unconjugated | |
| P21860 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| seribantumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| ERBB3/ HER3 | |
| 100 μg | |
| Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2065 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction