missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3/Her3 Antibody (SGP1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2692
This item is not returnable.
View return policy
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
Specifications
| ErbB3/Her3 | |
| Monoclonal | |
| 0.82 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| c-erb B3 protein from transfected human kidney fibroblasts purified by wheatgerm lectin affinity chromatography | |
| 0.1 mg | |
| Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2065 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF | |
| SGP1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction