Learn More
ERBB3 Monoclonal Antibody (OTI10D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809282
Encompass_Preferred
 View more versions of this product
Product Code. 17591344
  • € 540.87 / 100µL
Description
ErbB3 (c-erbB-3/HER-3) is a member of the type I family of growth factor receptors and binds to ligands in the heregulin family. ErbB3 is over-expressed in a variety of tumors in the prostate, bladder, breast, stomach, pancreas, and colon. Heregulin and EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents. The ErbB3 gene is located on the long arm of human chromosome 12 (12q13) and is transcribed into a 6.2kb mRNA which is translated into a 160/180kDa glycoprotein. The ErbB3 gene encodes a transmembrane receptor tyrosine kinase due to alternative splicing and forms heterodimers with other EGF receptor family members that have kinase activity. Heterodimerization leads to the activation of pathways that lead to cell proliferation or differentiation. Alternate transcriptional splice variants encoding different isoforms of ErbB3 have been characterized. One ErbB3 isoform lacks the intermembrane region, is secreted outside the cell, and acts to modulate the activity of the membrane-bound form. Additional splice variants have also been reported, but they have not been thoroughly characterized.
Specifications
Specifications
ERBB3
Monoclonal
Unconjugated
ERBB3
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
Mouse
Affinity Chromatography
RUO
2065
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI10D10
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P21860
ERBB3
Human recombit protein fragment corresponding to amino acids 983-1342 of human ERBB3 produced in E.coli.