missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERBB3 Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803827S
This item is not returnable.
View return policy
Description
ERBB3 Monoclonal antibody specifically detects ERBB3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ERBB3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08962 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI1A6 | |
| Unconjugated | |
| CD63 | |
| avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| Full length human recombinant protein of human CD63 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction