missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (50025), Alexa Fluor 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB38332AF405
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Western Blot.
Specifications
| CD14 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD14 | |
| Partial recombinant human CD14 protein (aa20-352) | |
| 100 μL | |
| Adaptive Immunity, Apoptosis, Asthma, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| 929 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 50025 | |
| Western Blot | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction