missing translation for 'onlineSavingsMsg'
Learn More

ErbB4/Her4 Antibody (HFR1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-2662B

Product Code. 18035564

  • € 456.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ErbB4/Her4
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
ERBB4
Synthetic peptide sequence RSTLQHPDYLQEYST from the cytoplasmic domain of the protein (residues 1249-1264.)
0.1 mL
Angiogenesis, Cancer, Protein Kinase, Tumor Suppressors
2066
Store at 4C in the dark.
IgG2b
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
HFR1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian)
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.