Learn More
ErbB4/Her4 Antibody (HFR1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2662MFV450
Product Code. 18724962
  • € 552.00 / 0.10mL
Description
ErbB4/Her4 Monoclonal antibody specifically detects ErbB4/Her4 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse
IgG2b