missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ErbB4 Recombinant Rabbit Monoclonal Antibody (8E5D0)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA535228

Product Code. 17125893

  • € 488.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DEYVNEPLYL NTFANTLGKA EYLKNNILSM PEKAKKAFDN PDYWNHSLPP RSTLQHPDYL QEYSTKYFYK QNGRIRPIVA ENPEYLSEFS LKPGTVLPPP PYRHRNTVV.

Oncogene ERBB4 (HER4) is a member of the type I receptor protein tyrosine subfamily that includes EGFR, ERBB2, and ERBB3. ERBB4 binds and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. ErbB 4 is activated by neuregulins and NTAK (neural and thymus derived activator for ErbB kinases). ErbB 4 is a receptor for heregulin and is capable of mediating HGL-stimulated tyrosine phosphorylation of itself. Like other protein kinases, ErbB 4 mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. ErbB 4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDAMB-330 and MDA-MB-361. Expression of ErbB 4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3. ErbB 4 levels have been found to be elevated in certain human tumor cell lines suggesting that it may play a role in some malignancies.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ErbB4
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q61527, Q62956
ERBB4
A synthetic peptide corresponding to a sequence within amino acids 1200-1308 of human ErbB4/HER4 (Q15303).
100 μL
Primary
Mouse, Rat
Antibody
IgG
ELISA, Western Blot
8E5D0
Unconjugated
ERBB4
4ICD; ALS19; avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4; avian erythroblastosis oncogene B 4; c-erbB-4; erb-b2 receptor tyrosine kinase 4; Erbb4; ERBB4 intracellular domain; ERBB4 transcript variant I12DEL; ERBB4 transcript variant I20DEL; Her4; human epidermal growth factor receptor 4; Mer4; OTTHUMP00000209432; p180erbB4; proto-oncogene-like protein c-ErbB-4; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-4; s80HER4; Tyro-2; Tyrosine kinase-type cell surface receptor HER4; v-erb-a erythroblastic leukemia viral oncogene homolog 4; v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian); v-erb-b2 avian erythroblastic leukemia viral oncogene 4; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4
Rabbit
Affinity chromatography
RUO
13869, 59323
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.