missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Antibody (8F1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-704AF594
This item is not returnable.
View return policy
Description
ERCC1 Monoclonal antibody specifically detects ERCC1 in Human, Rat, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ERCC1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Full length recombinant human ERCC1. [UniProt# P07992] | |
| 0.1 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2067 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8F1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat, Chinese Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction