missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Antibody (ERCC1/2318), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08903F
This item is not returnable.
View return policy
Description
ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Western Blot, ELISA, Protein Array
Specifications
| ERCC1 | |
| Monoclonal | |
| FITC | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Recombinant fragment (around aa 191-281) of human ERCC1 protein (exact sequence is proprietary) (Uniprot: P07992) | |
| 100 μg | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2067 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Protein Array | |
| ERCC1/2318 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction