missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Antibody (OTI2H1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70685FR
This item is not returnable.
View return policy
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ERCC1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| ERCC1 | |
| Full length recombinant protein of human ERCC1 (NP_001974) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2H1 | |
| Western Blot, Immunohistochemistry | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Mouse | |
| Affinity Purified | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2067.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction