missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Monoclonal Antibody (OTI2D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501181S
This item is not returnable.
View return policy
Description
ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ERCC1 | |
| Monoclonal | |
| 0.67 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00746 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4833 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2D8 | |
| Unconjugated | |
| NME4 | |
| COFS4; DNA excision repair protein ERCC-1; ERCC excision repair 1, endonuclease non-catalytic subunit; Ercc1; Ercc-1; excision repair cross-complementation group 1; excision repair cross-complementing 1; excision repair cross-complementing rodent repair deficiency, complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); hypothetical protein LOC405807; RAD10; UV20; zgc:77511 | |
| Full length human recombinant protein of human NME4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction