Learn More
Invitrogen™ ERCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592374
Description
Positive Samples: Raji, MCF-7, PC-3, HepG2, HeLa, Mouse liver, Mouse brain; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: MDPGKDKEGV PQPSGPPARK KFVIPLDEDE VPPGVAKPLF RSTQSLPTVD TSAQAAPQTY AEYAISQPLE GAGATCPTGS EPLAGETPNQ ALKPGAKSNS IIVSPRQRGN PVLKFVRNVP WEFGDVIPDY VLGQSTCALF LSLRYHNLHP DYIHGRLQSL GKNFALRVLL VQVDVKDPQQ ALKELAKMCI LADCTLILAW SPEEAGRYLE TYKAYEQKPA DLLMEKLEQD FVSRVTECLT TVKSVNKTDS QTLLTTFGSL EQLIAASRED LALCPGLGPQ KVRALGKNPR SWGKERAPNK HNLRPQSFKV KKEPKTRHSG FRL.
ERCC1 is a non-catalytic component of a structure-specific DNA repair endonuclease responsible for the 5'-incision during DNA repair. It is responsible, in conjunction with SLX4, for the first step in the repair of interstrand cross-links (ICL). It participates in the processing of anaphase bridge-generating DNA structures, which consist in incompletely processed DNA lesions arising during S or G2 phase, and can result in cytokinesis failure. ERCC1 is also required for homology-directed repair (HDR) of DNA double-strand breaks, in conjunction with SLX4. (Uniprot).
Specifications
| ERCC1 | |
| Polyclonal | |
| Unconjugated | |
| ERCC1 | |
| COFS4; DNA excision repair protein ERCC-1; ERCC excision repair 1, endonuclease non-catalytic subunit; Ercc1; Ercc-1; excision repair cross-complementation group 1; excision repair cross-complementing 1; excision repair cross-complementing rodent repair deficiency, complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); hypothetical protein LOC405807; RAD10; UV20; zgc:77511 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13870, 2067, 292673 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07903, P07992 | |
| ERCC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human ERCC1 (NP_9737301). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.