missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC6L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | ERCC6L |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637622
|
Novus Biologicals
NBP2-97609-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692932
|
Novus Biologicals
NBP2-97609-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERCC6L Polyclonal antibody specifically detects ERCC6L in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ERCC6L | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.0) | |
| 54821 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP-dependent helicase ERCC6-like, DNA excision repair protein ERCC-6-like, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 6-like, excision repair protein ERCC6-like, FLJ20105, MGC131695, PICHexcision repair cross-complementing rodent repair deficiency complementationgroup 6 - like, PLK1-interacting checkpoint helicase, SNF2/RAD54 family protein, Tumor antigen BJ-HCC-15 | |
| Produced in rabbits immunized with E. coli-derived Human ERCC6L fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title