missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ERCC8 Recombinant Rabbit Monoclonal Antibody (23GB5905), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

€ 195.00 - € 468.00

Specifications

Antigen ERCC8
Clone 23GB5905
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30087979
GREENER_CHOICE
View Documents
Invitrogen™
MA552946
100 μL
€ 468.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089596
View Documents
Invitrogen™
MA552947
20 μL
€ 195.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ERCC8 Recombinant Monoclonal antibody specifically detects ERCC8 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

ERCC8
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
Q13216
1161
IgG
Affinity chromatography
Antibody
23GB5905
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
2410022P04Rik; 2810431L23Rik; 4631412O06Rik; B130065P18Rik; Ckn1; Cockayne syndrome 1 (classical); Cockayne syndrome 1 homolog; cockayne syndrome WD repeat protein CSA; Cockayne syndrome WD repeat protein CSA homolog; Cockayne syndrome WD-repeat protein CSA; CSA; DNA excision repair protein ERCC-8; ERCC excision repair 8, CSA ubiquitin ligase complex subunit; Ercc8; excision repair cross-complementation group 8; excision repair cross-complementing rodent repair deficiency complementation group 8; excision repair cross-complementing rodent repair deficiency, complementation group 8; excision repaiross-complementing rodent repair deficiency, complementation group 8; UVSS2
A synthesized peptide derived from human ERCC8 (351-396AA).
Primary
-20°C
ERCC8
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado