missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eRF1 Antibody (OTI2E3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70692AF488
This item is not returnable.
View return policy
Description
eRF1 Monoclonal specifically detects eRF1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| eRF1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ETF1 | |
| Full length recombinant protein of human eRF1 (NP_004721) produced in HEK293T cells. | |
| 0.1 mL | |
| 2107.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI2E3 | |
| Western Blot, Flow Cytometry | |
| D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction