missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERG Antibody (ERG/6980), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20947H
This item is not returnable.
View return policy
Description
ERG Monoclonal antibody specifically detects ERG in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ERG | |
| Monoclonal | |
| HRP | |
| transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian) | |
| Recombinant fragment (around aa450 to 479) (C-terminus) of human ERG (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 2078 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| ERG/6980 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction