missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERG Antibody (Jackson Foundation patent anti-ERG), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28267CL1
This item is not returnable.
View return policy
Description
ERG Monoclonal antibody specifically detects ERG in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ERG | |
| Monoclonal | |
| CoraFluor 1 | |
| transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian) | |
| ERG | |
| 0.1 mL | |
| Cancer | |
| 2078 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Jackson Foundation patent anti-ERG | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction