missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFS1 Mouse anti-Human, Clone: OTI2H5, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805491S
This item is not returnable.
View return policy
Description
NFS1 Monoclonal antibody specifically detects NFS1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NFS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y697 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9054 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2H5 | |
| Unconjugated | |
| NFS1 | |
| AA987187; cysteine desulfurase, mitochondrial; Cysteine desulfurase, mitochondrial-like protein; HUSSY-08; IscS; LOW QUALITY PROTEIN: cysteine desulfurase, mitochondrial; m-Nfs1; m-Nfsl; Nfs1; NFS1 cysteine desulfurase; NFS1 nitrogen fixation 1 homolog; NFS1 nitrogen fixation 1 homolog (S. cerevisiae); NFS1, cysteine desulfurase; Nifs; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene 1 (S. cerevisiae); nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein | |
| Human recombinant protein fragment corresponding to amino acids 1-299 of human NFS1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction