missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERG Monoclonal Antibody (OTI5F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506077S
This item is not returnable.
View return policy
Description
ERG Monoclonal antibody specifically detects ERG in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| ERG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11308 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2078 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5F12 | |
| Unconjugated | |
| Erg | |
| avian erythroblastosis virus E-26 (v-ets) oncogene related; D030036I24Rik; ERG; ERG, ETS transcription factor; ERG/EWS fusion gene; ERG/FUS fusion gene; ERG/TMPSSR2 fusion gene; ERG1; Erg-3; erythroblast transformation-specific transcription factor ERG variant 10; ETS transcription factor ERG; ets-related; H Kv11.3; HGNC:3446; KCNH2; Oncogene ERG; OTTHUMP00000115970; OTTHUMP00000208036; p55; TMPRSS2/ERG fusion; transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); transcriptional regulator ERG; LOW QUALITY PROTEIN: transcriptional regulator ERG; Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like | |
| Full length human recombinant protein of human ERG produced in HEk293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction