Learn More
SENP8 Monoclonal Antibody (OTI10C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805442S
Encompass_Preferred
 View more versions of this product
Product Code. 17535373
  • € 242.00 / 30µL
Description
SENP8 Monoclonal antibody specifically detects SENP8 in Human samples. It is validated for Western Blot
Specifications
Specifications
SENP8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42771
Mouse
Affinity chromatography
RUO
1029
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI10C11
Unconjugated
CDKN2A
9130010J17Rik; AU020827; DEN1; deneddylase 1; deneddylase-1; FKSG8; NEDD8 specific-protease cysteine 2; NEDD8-specific protease 1; Nedp1; protease, cysteine 2; protease, cysteine, 2 (NEDD8 specific); PRSC2; SENP8; sentrin/SUMO-specific protease SENP8; Sentrin-specific protease 8; SUMO sentrin specific protease family member 8; SUMO/sentrin peptidase family member, NEDD8 specific; SUMO/sentrin specific peptidase 8; SUMO/sentrin specific peptidase family member 8; SUMO/sentrin specific protease 8; SUMO/sentrin specific protease family member 8; Sumo1/sentrin/SMT3 specific peptidase 8
Full length human recombinant protein of human CDKN2A produced in E.coli.