missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERG Mouse anti-Human, Clone: OTI8A9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506201
This item is not returnable.
View return policy
Description
Transcriptional regulator. May participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.Specifications
| ERG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11308 | |
| ERG | |
| Full length human recombit protein of human ERG produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A9 | |
| Unconjugated | |
| ERG | |
| erg-3, p55 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2078 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction