missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERI1 Polyclonal Antibody

ERI1 Polyclonal Antibody
Brand: Invitrogen PA517141
This item is not returnable.
View return policy
Description
ERI1 Polyclonal Antibody for Western Blot, IP
Specifications
| ERI1 | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q5FVR4, Q7TMF2 | |
| ERI1 | |
| Synthetic peptide corresponding to the sequence of mouse THEX1. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ERI1 | |
| 3 ' exoribonuclease; 3' exoribonuclease; 3110010F15Rik; 3'-5' exonuclease ERI1; 3'-5' exoribonuclease 1; 3'EXO; 3'HEXO; enhanced RNAi protein 1; enhanced RNAi three prime mRNA exonuclease homolog 1; ERI1; eri-1; Eri-1 homolog; exoribonuclease 1; HEXO; histone mRNA 3' end-specific exonuclease; histone mRNA 3'-end-specific exoribonuclease; Histone mRNA 3'-exonuclease 1; protei; protein 3'hExo; RGD1308378; Thex1; three prime histone mRNA exonuclease 1; three prime mRNA exonuclease 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361159, 67276 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction