missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ERI1 Polyclonal Antibody

Product Code. 15731022 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15731022 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15731022 Supplier Invitrogen™ Supplier No. PA563244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ELRAKLSEFK LETRGVKDVL KKRLKNYYKK QKLMLKESNF ADSYYDYICI IDFEATCEE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

RNA exonuclease that binds to the 3'-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. A 2' and 3'-hydroxyl groups at the last nucleotide of the histone 3'-end is required for efficient degradation of RNA substrates. Also able to degrade the 3'-overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi). Requires for binding the 5'-ACCCA-3' sequence present in stem-loop structure. Able to bind other mRNAs. Required for 5.8S rRNA 3'-end processing. Also binds to 5.8s ribosomal RNA. Binds with high affinity to the stem-loop structure of replication-dependent histone pre-mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERI1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERI1
Gene Accession No. Q8IV48
Gene Alias 3 ' exoribonuclease; 3' exoribonuclease; 3110010F15Rik; 3'-5' exonuclease ERI1; 3'-5' exoribonuclease 1; 3'EXO; 3'HEXO; enhanced RNAi protein 1; enhanced RNAi three prime mRNA exonuclease homolog 1; ERI1; eri-1; Eri-1 homolog; exoribonuclease 1; HEXO; histone mRNA 3' end-specific exonuclease; histone mRNA 3'-end-specific exoribonuclease; Histone mRNA 3'-exonuclease 1; protei; protein 3'hExo; RGD1308378; Thex1; three prime histone mRNA exonuclease 1; three prime mRNA exonuclease 1
Gene Symbols ERI1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERI1. Recombinant protein control fragment (Product #RP-103246).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90459
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.