missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05652-100ul
This item is not returnable.
View return policy
Description
ERK1/2 Polyclonal antibody specifically detects ERK1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ERK1/2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ERK1/2ERK1/2 (P27361/P28482). FLTEYVATRWYRAPEIMLNSKGYTKSIDIWSVGCILAEMLSNRPIFPGKHYLDQLNHILGILGSPSQEDLNCIINMKARNYLQSLPSKTKVAWAKLFPKSD/LNSKGYTKSIDIWSVGCILAEMLSNRPIFPGKHYLDQLNHILGILGSPSQEDLNCIINLKARNYLLSLPHKNKVPWNRLFPNADSKALDLLDKMLTFNPHK | |
| 100 μg | |
| Alzheimers Research, Angiogenesis, Apoptosis, Breast Cancer, Cancer, Caspases, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Neurodegeneration, Protein Kinase, Signal Transduction | |
| 5595 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction