missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ERK1/2 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856355
|
Proteintech
51068-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846355
|
Proteintech
51068-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can recognize both ERK1 and ERK2 with the molecular mass of 38-43 kDa.
MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. The p44/42 MAPK (Erk1/2) are members of mitogen-activated kinases (MAPKs) of serine/threonine protein kinases. Phospho ERK1/2 can be activated by a range of extracellular stimuli, such as mitogen, growth factors, neurotransmitters, chemokines, and cytokines, through receptor tyrosine kinases (RTK), G protein-coupled receptors (GPCRs), or protein kinase C (PKC). Upstream MEK1 and MEK2 activate p44 and p42 (phospho ERK1/2) through phosphorylation of activation loop residues Thr202/Tyr204 and Thr185/Tyr187, respectively. p44/42 (ERK1/2) are negatively regulated by a family of dual-specificity (Thr/Tyr) MAPK phosphatases, known as DUSPs or MKPs, along with MEK inhibitors, such as U0126 and PD98059.Specifications
| ERK1/2 | |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P21708, P27361, P28482, P63085, P63086, Q63844 | |
| 116590, 26413, 26417, 50689, 5594, 5595 | |
| Peptide | |
| Primary | |
| -20°C | |
| MAPK1 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ERK, ERK 2, ERK1/2, ERK2, ERT1, MAP kinase 1, MAP kinase 2, MAP kinase isoform p42, MAPK 1, MAPK 2, MAPK1, MAPK2, p40, p41, p41mapk, p42 MAPK, P42MAPK, PRKM1, PRKM2 | |
| Mapk1, Mapk3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title