missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/ERK2 [p Thr202, p Thr185, p Tyr187, p Tyr204] Antibody (269434), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB1018CL1
This item is not returnable.
View return policy
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human,Mouse,Rat samples. It is validated for Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| ERK1/ERK2 | |
| Monoclonal | |
| CoraFluor 1 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 269434 | |
| PBS | |
| Phosphopeptide containing ERK1 T202/Y204 site | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction