missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/ERK2 [p Tyr187, p Tyr204, p Thr185, p Thr202] Antibody (269434) [DyLight 594], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB1018M
This item is not returnable.
View return policy
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| ERK1/ERK2 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Phosphopeptide containing ERK1 T202/Y204 site | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 269434 | |
| Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction