Learn More
ERK1/ERK2, p Tyr204 Antibody (SR1718), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21522-100ul
 View more versions of this product
Product Code. 18678854
  • € 449.00 / 100µL
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ERK1/ERK2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
A synthesized peptide derived from human phospho-ERK1 [Tyr204]/ERK2 [Tyr187] (Uniprot #: P27361, P28482)
100 μg
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
IgG
Western Blot, Immunoprecipitation
SR1718
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Rabbit
Affinity purified
RUO
Human, Mouse, Rat