missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 277.00 - € 456.00
Specifications
| Antigen | ERK2 |
|---|---|
| Dilution | Western Blot 1:100-1:250, Simple Western 1:5, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453712
|
Novus Biologicals
NBP1-87943-25ul |
25 μL |
€ 277.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18411882
|
Novus Biologicals
NBP1-87943 |
0.1 mL |
€ 456.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERK2 Polyclonal antibody specifically detects ERK2 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ERK2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Protein Kinase, Signal Transduction, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol | |
| 5594 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 1:100-1:250, Simple Western 1:5, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YIVQDLMETDLYKLLKTQHLSNDHICY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title