Learn More
ERK2 Antibody (OTI6E5), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71233AF350
Code produit. 18165453
  • € 751.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
ERK2 Monoclonal specifically detects ERK2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
ERK2
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
MAPK1
Full length recombinant protein of human ERK2 (NP_620407) produced in HEK293T cells.
Primary
Human, Mouse, Rat, Canine, Monkey
IgG1
Western Blot, Immunohistochemistry
OTI6E5
Western Blot, Immunohistochemistry
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Mouse
0.1 mL
5594
Store at 4°C in the dark.