missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK5/BMK1 Antibody (OTI3H2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71243AF488
This item is not returnable.
View return policy
Description
ERK5/BMK1 Monoclonal specifically detects ERK5/BMK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| ERK5/BMK1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAPK7 | |
| Full length recombinant protein of human ERK5/BMK1 (NP_002740) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI3H2 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Big MAP kinase 1, BMK-1, BMK1 kinase, BMK1EC 2.7.11.24, EC 2.7.11, ERK-5, ERK5MAPK 7, Extracellular signal-regulated kinase 5, extracellular-signal-regulated kinase 5, MAP kinase 7, mitogen-activated protein kinase 7, PRKM7ERK4 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| 5598.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction