Learn More
Fisherbrand™ Erlenmeyer Flask Clamp for Fisherbrand™ Shaking Incubators
Erlenmeyer Flask Clamp for use with Fisherbrand™ Shaking Incubators with magnetic platform.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 17749199
  • € 129.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • Magnetic attachment
  • Toolless exchange
  • Holds flask
Specifications
Specifications
3 L Erlenmeyer Flask
Fisherbrand Shaking Incubator