missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERMP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532759
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%) Mouse, Rat, Hamster, Rabbit, Pig (93%).
Within the ovary, required for the organization of somatic cells and oocytes into discrete follicular structures.
Specifications
| ERMP1 | |
| Polyclonal | |
| Unconjugated | |
| ERMP1 | |
| aminopeptidase Fxna; bA207C16.3; bA207C16.3 (novel protein similar to predicted yeast, plant and worm proteins); D19Ertd410e; D19Wsu12e; Endoplasmic reticulum metallopeptidase 1; Ermp1; Felix-ina; Fxna; KIAA1815; mKIAA1815; putative aminopeptidase Fxna | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q7Z2K6 | |
| ERMP1 | |
| Synthetic 14 amino acid peptide from internal region of human ERMP1. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction