missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERO1L Antibody (V23P3C9*H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 351.00
Specifications
| Antigen | ERO1L |
|---|---|
| Clone | V23P3C9*H2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624788
|
Novus Biologicals
NBP2-50388-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656838
|
Novus Biologicals
NBP2-50388 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERO1L Monoclonal antibody specifically detects ERO1L in Human, Mouse, Rat, Porcine, Canine, Equine, Sheep samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| ERO1L | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction | |
| PBS | |
| 30001 | |
| IgG1 κ | |
| Protein A purified |
| V23P3C9*H2 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Canine | |
| EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha | |
| Peptide Sequence - KLIANMPESG (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), sheep (Ovis aries), bovine (Bos taurus), canine (Canis familiaris), porcine (Sus scrofa) and equine (Equus caballus) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title